missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diacetoxy-2-butyne 95.0+%, TCI America™

Catalog No. B208725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B208725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B208725G Supplier TCI America Supplier No. B208725G

Chemical Identifiers

CAS 1573-17-7
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.16
MDL Number MFCD00014982
InChI Key TVIMIQVBDDNCCR-UHFFFAOYSA-N
Synonym 1,4-diacetoxy-2-butyne, but-2-yne-1,4-diyl diacetate, butynediol diacetate, 2-butyne-1,4-diol diacetate, 2-butyne-1,4-diol, diacetate, 2-butyne-1,4-diol, 1,4-diacetate, 4-acetyloxy but-2-yn-1-yl acetate, acmc-209dfx, 2-butyne-1, diacetate, bis acetoxymethyl acetylene
PubChem CID 74081
IUPAC Name 4-(acetyloxy)but-2-yn-1-yl acetate
SMILES CC(=O)OCC#CCOC(C)=O

Specifications

Color White-Yellow
Boiling Point 235°C
Quantity 25 g
Formula Weight 170.16
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,4-Diacetoxy-2-butyne