missing translation for 'onlineSavingsMsg'
Learn More

(-)-1,4-Di-O-tosyl-2,3-O-isopropylidene-L-threitol 98.0+%, TCI America™

Catalog No. D16231G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16231G 1 g
D16235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16231G Supplier TCI America Supplier No. D16231G

Chemical Identifiers

CAS 37002-45-2
Molecular Formula C21H26O8S2
Molecular Weight (g/mol) 470.55
MDL Number MFCD00003212,MFCD00003212,MFCD00063235
InChI Key KPFDKWNWYAXRNJ-UHFFFAOYNA-N
Synonym 2,2-dimethyl-4,5-bis tosyloxymethyl-1,3-dioxolane, 2,2-dimethyl-5-4-methylphenyl sulfonyloxymethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate, --2,3-o-isopropylidene-1,4-di-o-tosyl-l-threitol, 1,3-dioxolane-4,5-dimethanol, 2,2-dimethyl-, bis 4-methylbenzenesulfonate, s,s---1,4-di-o-tosyl-2,3-o-isopropy lidene-l-threitol, --1,4-ditosyl-2,3,o-isopropylidene-l-threitol, --1,4-ditosyl-2,3-o-isopropylidene-l-threitol, --1,4-di-o-tosyl-2,3-o-isopropylidene-threitol, l-o-isopropylidene-2,3-dihydroxy-1,4-bis p-tosyl butane, 4s,5s---1,4-di-o-tosyl-2,3-o-isopropylidene-l-threitol
PubChem CID 275953
IUPAC Name (2,2-dimethyl-5-{[(4-methylbenzenesulfonyl)oxy]methyl}-1,3-dioxolan-4-yl)methyl 4-methylbenzene-1-sulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC1OC(C)(C)OC1COS(=O)(=O)C1=CC=C(C)C=C1

Specifications

Melting Point 93°C
Color White-Yellow
Quantity 1 g
Formula Weight 470.55
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (-)-1,4-Di-O-tosyl-2,3-O-isopropylidene-L-threitol