missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanedione Monoethyleneketal 98.0+%, TCI America™

Catalog No. p-7124899
Change view
Click to view available options
Quantity:
100 g
25 g
5 g
Catalog No. Quantity
C12925G 5 g
C129225G 25 g
C1292100G 100 g
3 options

Catalog No. C12925G

Supplier: TCI America C12925G

Only null left

Chemical Identifiers

CAS 4746-97-8
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.181
MDL Number MFCD00010214
InChI Key VKRKCBWIVLSRBJ-UHFFFAOYSA-N
Synonym 1,4-dioxaspiro 4.5 decan-8-one, 1,4-cyclohexanedione monoethylene acetal, 1,4-cyclohexanedione monoethylene ketal, 1,4-cyclohexanedione monoethyleneacetal, 1,4-dioxa-spiro 4.5 decan-8-one, 1,4-cyclohexanedione monoethyleneketal, cyclohexane-1,4-dione monoethylene ketal, 1,4-dioxaspiro 4.5 decane-8-one
PubChem CID 567415
IUPAC Name 1,4-dioxaspiro[4.5]decan-8-one
SMILES C1CC2(CCC1=O)OCCO2

Specifications

Melting Point 73°C
Quantity 5 g
Formula Weight 156.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Cyclohexanedione Monoethyleneketal