missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanedione monoethylene acetal, 97%

Catalog No. AAA1040806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1040806 5 g
AAA1040814 25 g
AAA1040822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1040806 Supplier Thermo Scientific Chemicals Supplier No. A1040806

CAS: 4746-97-8 | C8H12O3 | 156.181 g/mol

1,4-Cyclohexanedione is used in the preparation of series of potent analgesic compounds. 1,4-Cyclohexanedione is also used as a building block in the synthesis of tritium labelled probes for the autoradiography study of the dopamine reuptake complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Cyclohexanedione is used in the preparation of series of potent analgesic compounds. 1,4-Cyclohexanedione is also used as a building block in the synthesis of tritium labelled probes for the autoradiography study of the dopamine reuptake complex.

Solubility
Soluble in chloroform & methanol.

Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Incompatible with strong oxidizing agents and strong acids. Ensure adequate ventilation during use.

Chemical Identifiers

CAS 4746-97-8
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.181
MDL Number MFCD00010214
InChI Key VKRKCBWIVLSRBJ-UHFFFAOYSA-N
Synonym 1,4-dioxaspiro 4.5 decan-8-one, 1,4-cyclohexanedione monoethylene acetal, 1,4-cyclohexanedione monoethylene ketal, 1,4-cyclohexanedione monoethyleneacetal, 1,4-dioxa-spiro 4.5 decan-8-one, 1,4-cyclohexanedione monoethyleneketal, cyclohexane-1,4-dione monoethylene ketal, 1,4-dioxaspiro 4.5 decane-8-one
PubChem CID 567415
IUPAC Name 1,4-dioxaspiro[4.5]decan-8-one
SMILES C1CC2(CCC1=O)OCCO2

Specifications

Melting Point 72°C to 75°C
Quantity 5 g
Beilstein 117631
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform & methanol.
Formula Weight 156.18
Percent Purity 97%
Chemical Name or Material 1,4-Cyclohexanedione monoethylene acetal
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only