missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanedione Bis(ethyleneketal) 99.0+%, TCI America™

Catalog No. C154425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C154425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C154425G Supplier TCI America Supplier No. C154425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 183-97-1
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.234
MDL Number MFCD00010851
InChI Key YSMVSEYPOBXSOK-UHFFFAOYSA-N
Synonym 1,4-cyclohexanedione bis ethylene ketal, 1,4,9,12-tetraoxadispiro 4.2.4.2 tetradecane, 1,4-cyclohexanedione bis ethylene acetal, 1,4,9,12-tetraoxadispiro 4,2,4,2 tetradecane, 1,4-cyclohexanedione bis ethyleneketal, 1,4,9,12-tetraoxadispiro 4.2.4^ 8 .2^ 5 tetradecane, 1,4,9,12-tetraoxadispiro 4.2.4?.2? tetradecane, maybridge1_006651, 1,4-cyclohexanedione ethyleneketal
PubChem CID 135986
IUPAC Name 1,4,9,12-tetraoxadispiro[4.2.4^{8}.2^{5}]tetradecane
SMILES C1CC2(CCC13OCCO3)OCCO2

Specifications

Melting Point 78°C
Color White-Yellow
Quantity 25 g
Formula Weight 200.23
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Cyclohexanedione Bis(ethyleneketal)