missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanedione bis(ethylene acetal), 99%

Catalog No. AAB2078430
Change view
Click to view available options
Quantity:
10 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2078430 250 g
AAB2078409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2078430 Supplier Thermo Scientific Chemicals Supplier No. B2078430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 183-97-1 | C10H16O4 | 200.234 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 183-97-1
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.234
MDL Number MFCD00010851
InChI Key YSMVSEYPOBXSOK-UHFFFAOYSA-N
Synonym 1,4-cyclohexanedione bis ethylene ketal, 1,4,9,12-tetraoxadispiro 4.2.4.2 tetradecane, 1,4-cyclohexanedione bis ethylene acetal, 1,4,9,12-tetraoxadispiro 4,2,4,2 tetradecane, 1,4-cyclohexanedione bis ethyleneketal, 1,4,9,12-tetraoxadispiro 4.2.4^ 8 .2^ 5 tetradecane, 1,4,9,12-tetraoxadispiro 4.2.4?.2? tetradecane, maybridge1_006651, 1,4-cyclohexanedione ethyleneketal
PubChem CID 135986
IUPAC Name 1,4,9,12-tetraoxadispiro[4.2.4^{8}.2^{5}]tetradecane
SMILES C1CC2(CCC13OCCO3)OCCO2

Specifications

Melting Point 79°C to 80°C
Boiling Point 75°C to 85°C (0.5 mmHg)
Quantity 250 g
Formula Weight 200.23
Percent Purity 99%
Chemical Name or Material 1,4-Cyclohexanedione bis(ethylene acetal)
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only