missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanedione, 98%

CAS: 637-88-7 | C6H8O2 | 112.128 g/mol

Supplier:  Thermo Scientific Chemicals A1110022

 View more versions of this product

Catalog No. AAA1110022


Only null left
Explore available promotions

Description

Description

1,4-Cyclohexanedione is used in the preparation of 1,4 benzoquinone and bromoorganics. It is also used to study the influence of visible light on the bromate-1,4-cyclohexanedione-ferroin oscillating reaction. It plays a vital role in pharmaceuticals, plant growth regulator and as a conducting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Cyclohexanedione is used in the preparation of 1,4 benzoquinone and bromoorganics. It is also used to study the influence of visible light on the bromate-1,4-cyclohexanedione-ferroin oscillating reaction. It plays a vital role in pharmaceuticals, plant growth regulator and as a conducting material.

Solubility
Soluble in methanol and ethanol.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

1,4-Cyclohexanedione
77°C to 78°C
132°C (269°F)
MFCD00001606
774152
Soluble in methanol and ethanol.
C1CC(=O)CCC1=O
112.128
CHEBI:28286
98%
637-88-7
132°C (20 mmHg)
C6H8O2
100 g
1,4-cyclohexanedione, tetrahydroquinone, 1,4-dioxocyclohexane, 1,4-cyclohexandione, 1,4 cyclohexane dione, 1,4-cyclohexane-dione, cyclohexan-1,4-dione, pubchem13686, cyclohexane-1,4-quinone, acmc-1b8ar
DCZFGQYXRKMVFG-UHFFFAOYSA-N
cyclohexane-1,4-dione
12511
112.13

Chemical Identifiers

637-88-7
112.128
DCZFGQYXRKMVFG-UHFFFAOYSA-N
12511
cyclohexane-1,4-dione
C6H8O2
MFCD00001606
1,4-cyclohexanedione, tetrahydroquinone, 1,4-dioxocyclohexane, 1,4-cyclohexandione, 1,4 cyclohexane dione, 1,4-cyclohexane-dione, cyclohexan-1,4-dione, pubchem13686, cyclohexane-1,4-quinone, acmc-1b8ar
CHEBI:28286
C1CC(=O)CCC1=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-306-0

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only