missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanediol (cis- and trans- mixture) 99.0+%, TCI America™

Catalog No. C0482500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0482500G 500 g
C048225G 25 g
C0482100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0482500G Supplier TCI America Supplier No. C0482500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 556-48-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00001448,MFCD00063612,MFCD00075462
InChI Key VKONPUDBRVKQLM-UHFFFAOYSA-N
Synonym 1,4-cyclohexanediol, cis-1,4-cyclohexanediol, trans-1,4-cyclohexanediol, trans-cyclohexane-1,4-diol, quinitol, 1,4-cyclohexanediol, trans, cis-cyclohexane-1,4-diol, 1,4-cyclohexanediol, cis, trans-1,4-dihydroxycyclohexane, cis-1,4-dihydroxycyclohexane
PubChem CID 11162
IUPAC Name cyclohexane-1,4-diol
SMILES OC1CCC(O)CC1

Specifications

Melting Point 101°C
Color White
Boiling Point 132°C
Quantity 500 g
Formula Weight 116.16
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Cyclohexanediol (cis- and trans- mixture)
EINECSNumber (3)-2310
TSCA No