missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanediol, cis + trans, 98%

Catalog No. p-7025112
Change view
Click to view available options
Quantity:
25 g
100 g
Catalog No. Quantity
AAB2044614 25 g
AAB2044622 100 g
2 options

Catalog No. AAB2044614

Supplier: Thermo Scientific Chemicals B2044614

Only null left

CAS: 556-48-9 | C6H12O2 | 116.16 g/mol

1,4-Cyclohexanediol, cis + trans, is used as a chemical, organic, pharmaceutical intermediate. It is also used as an important intermediate for manufacturing of anti-Cancer and anti-Infective Drugs - dihydroartemisinin and dihydroartemisitene dimers. It is also used in co polyester resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Cyclohexanediol, cis + trans, is used as a chemical, organic, pharmaceutical intermediate. It is also used as an important intermediate for manufacturing of anti-Cancer and anti-Infective Drugs - dihydroartemisinin and dihydroartemisitene dimers. It is also used in co polyester resins.

Solubility
It is highly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acid chlorides, acid anhydrides, strong acids.

Chemical Identifiers

CAS 556-48-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00001448,MFCD00063612,MFCD00075462
InChI Key VKONPUDBRVKQLM-UHFFFAOYSA-N
Synonym 1,4-cyclohexanediol, cis-1,4-cyclohexanediol, trans-1,4-cyclohexanediol, trans-cyclohexane-1,4-diol, quinitol, 1,4-cyclohexanediol, trans, cis-cyclohexane-1,4-diol, 1,4-cyclohexanediol, cis, trans-1,4-dihydroxycyclohexane, cis-1,4-dihydroxycyclohexane
PubChem CID 11162
IUPAC Name cyclohexane-1,4-diol
SMILES OC1CCC(O)CC1

Specifications

Melting Point 98°C to 103°C
Boiling Point 149°C to 151°C (20 mmHg)
Flash Point 65°C (149°F)
Quantity 25 g
Beilstein 2036658
Solubility Information It is highly soluble in water.
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material 1,4-Cyclohexanediol, cis + trans
EINECSNumber 209-126-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only