missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanediol, 99%, mixture of cis and trans

Catalog No. AC111150250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111150250 25 g Plastic bottle
AC111151000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC111150250 Supplier Thermo Scientific Chemicals Supplier No. 111150250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 556-48-9 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 556-48-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00001448,MFCD00063612,MFCD00075462
InChI Key VKONPUDBRVKQLM-UHFFFAOYSA-N
Synonym 1,4-cyclohexanediol, cis-1,4-cyclohexanediol, trans-1,4-cyclohexanediol, trans-cyclohexane-1,4-diol, quinitol, 1,4-cyclohexanediol, trans, cis-cyclohexane-1,4-diol, 1,4-cyclohexanediol, cis, trans-1,4-dihydroxycyclohexane, cis-1,4-dihydroxycyclohexane
PubChem CID 11162
IUPAC Name cyclohexane-1,4-diol
SMILES OC1CCC(O)CC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 98°C to 102°C
Boiling Point 150°C (20.0 mmHg)
Flash Point 65°C
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula C6H10(OH)2
Quantity 25 g
Beilstein 06, IV, 5209
Solubility Information Solubility in water: soluble
Formula Weight 116.16
Percent Purity 99%
Chemical Name or Material 1, 4-Cyclohexanediol, 99%
Show More Show Less
EINECSNumber 209-126-2

RUO – Research Use Only