missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanedimethanol (cis- and trans- mixture) 99.0+%, TCI America™

Catalog No. c047925g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
C047925G 25 g
C0479500G 500 g
2 options

Catalog No. C047925G

Supplier: TCI America C047925G

Only null left

Chemical Identifiers

CAS 105-08-8
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00001512,MFCD00066360
InChI Key YIMQCDZDWXUDCA-UHFFFAOYSA-N
Synonym 1,4-cyclohexanedimethanol, trans-1,4-cyclohexanedimethanol, chdm, 1,4-bis hydroxymethyl cyclohexane, rikabinol dm, cyclohexane-1,4-dimethanol, 1,4-chidm, 1,4-dimethylolcyclohexane, 4-hydroxymethyl cyclohexyl methanol, 1,4-cyclohexanedimethanol, cis
PubChem CID 7735
IUPAC Name [4-(hydroxymethyl)cyclohexyl]methanol
SMILES OCC1CCC(CO)CC1

Specifications

Color Colorless
Boiling Point 162°C
Quantity 25 g
Formula Weight 144.21
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Cyclohexanedimethanol (cis- and trans- mixture)
EINECSNumber (3)-2326
RTECSNumber GU9800000
TSCA Yes