missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Cyclohexanedicarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 172.18 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,4-Cyclohexanedicarboxylic Acid (cis- and trans- mixture) |
Safety and Handling
| EINECSNumber | (3)-2423&(3)-2446 |
| RTECSNumber | GU9060000 |
| TSCA | Yes |