missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Cyclohexanediamine (cis- and trans- mixture) 97.0+%, TCI America™
$46.60 - $134.21
Chemical Identifiers
| CAS | 3114-70-3 |
|---|---|
| Molecular Formula | C6H14N2 |
| Molecular Weight (g/mol) | 114.19 |
| MDL Number | MFCD00035862,MFCD00075174,MFCD08276323 |
| InChI Key | VKIRRGRTJUUZHS-UHFFFAOYSA-N |
| Synonym | 1,4-Diaminocyclohexane |
| PubChem CID | 18374 |
| IUPAC Name | cyclohexane-1,4-diamine |
| SMILES | NC1CCC(N)CC1 |
Chemical Identifiers
| 3114-70-3 | |
| 114.19 | |
| VKIRRGRTJUUZHS-UHFFFAOYSA-N | |
| 18374 | |
| NC1CCC(N)CC1 |
| C6H14N2 | |
| MFCD00035862,MFCD00075174,MFCD08276323 | |
| 1,4-Diaminocyclohexane | |
| cyclohexane-1,4-diamine |
Specifications
| 3114-70-3 | |
| 197°C | |
| MFCD00035862,MFCD00075174,MFCD08276323 | |
| 3259 | |
| VKIRRGRTJUUZHS-UHFFFAOYSA-N | |
| cyclohexane-1,4-diamine | |
| 18374 | |
| ≥97.0% (GC,T) | |
| 1,4-Cyclohexanediamine (cis- and trans- mixture) |
| White-Yellow | |
| C6H14N2 | |
| 5 mL | |
| 1,4-Diaminocyclohexane | |
| NC1CCC(N)CC1 | |
| 114.19 | |
| 114.19 | |
| Crystalline Lumps |
Safety and Handling
RTECSNumber : GU8750500
TSCA : No