missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Cyanophenyl)imidazole 98.0+%, TCI America™

Catalog No. C20455G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C20455G Supplier TCI America Supplier No. C20455G
Only null left

Chemical Identifiers

CAS 25372-03-6
Molecular Formula C10H7N3
Molecular Weight (g/mol) 169.19
MDL Number MFCD00127147
InChI Key GYFGZFJGMRRTTP-UHFFFAOYSA-N
Synonym 4-1h-imidazol-1-yl benzonitrile, 4-imidazol-1-yl benzonitrile, p-imidazol-1-ylbenzonitrile, 1-4-cyanophenyl imidazole, 4-1-imidazolyl benzonitrile, 4-1h-imidazol-1-yl benzenecarbonitrile, benzonitrile, p-imidazol-1-yl, 4-imidazolylbenzenecarbonitrile, benzonitrile, 4-1h-imidazol-1-yl, acmc-1cnga
PubChem CID 135744
IUPAC Name 4-(1H-imidazol-1-yl)benzonitrile
SMILES N#CC1=CC=C(C=C1)N1C=CN=C1

Specifications

Melting Point 154°C
Quantity 5 g
UN Number 3439
Formula Weight 169.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Cyanophenyl)imidazole