missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Cyanophenyl)guanidine 99.0+%, TCI America™

$50.33 - $106.25

Chemical Identifiers

CAS 5637-42-3
Molecular Formula C8H8N4
Molecular Weight (g/mol) 160.18
MDL Number MFCD16251323
InChI Key BTDGLZSKNFJBER-UHFFFAOYSA-N
Synonym 1-4-cyanophenyl guanidine, n-4-cyanophenyl guanidine, 2-4-cyanophenyl guanidine, 4-cyanophenyl guanidine, 4-cyanophenylguanidine
PubChem CID 17747914
IUPAC Name 2-(4-cyanophenyl)guanidine
SMILES C1=CC(=CC=C1C#N)N=C(N)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C33871G
View Documents
TCI America
C33871G
1 g N/A
Only null left
 
C33875G
View Documents
TCI America
C33875G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

5637-42-3
160.18
BTDGLZSKNFJBER-UHFFFAOYSA-N
17747914
C1=CC(=CC=C1C#N)N=C(N)N
C8H8N4
MFCD16251323
1-4-cyanophenyl guanidine, n-4-cyanophenyl guanidine, 2-4-cyanophenyl guanidine, 4-cyanophenyl guanidine, 4-cyanophenylguanidine
2-(4-cyanophenyl)guanidine

Specifications

5637-42-3
White-Yellow
MFCD16251323
1-4-cyanophenyl guanidine, n-4-cyanophenyl guanidine, 2-4-cyanophenyl guanidine, 4-cyanophenyl guanidine, 4-cyanophenylguanidine
C1=CC(=CC=C1C#N)N=C(N)N
160.18
160.18
Crystalline Powder
212°C
C8H8N4
1 g
BTDGLZSKNFJBER-UHFFFAOYSA-N
2-(4-cyanophenyl)guanidine
17747914
≥99.0% (HPLC,N)
1-(4-Cyanophenyl)guanidine
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'