missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cineole, 85%

Catalog No. p-7077598
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456060250 25 g
AC456061000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC456060250 Supplier Thermo Scientific Chemicals Supplier No. 456060250

CAS: 470-67-7 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 470-67-7
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
InChI Key RFFOTVCVTJUTAD-UHFFFAOYSA-N
Synonym 1,4-cineole, isocineole, 1,4-cineol, 1,4-epoxy-p-menthane, isocineple, 1,4-cineole natural, p-menthane, 1,4-epoxy, unii-b55jtu839b, fema no. 3658
PubChem CID 10106
ChEBI CHEBI:80788
IUPAC Name 1-methyl-4-propan-2-yl-7-oxabicyclo[2.2.1]heptane
SMILES CC(C)C12CCC(O1)(CC2)C

Specifications

Melting Point -46.0°C
Density 0.8870g/mL
Boiling Point 65°C (21.0 hPa)
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 80% min. (GC)
Quantity 25 g
Specific Gravity 0.887
Formula Weight 154.25
Percent Purity 85%
Chemical Name or Material 1,4-Cineole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.
EINECSNumber 207-428-9

RUO – Research Use Only