Learn More
1-(4-Chlorophenyl)ethylamine, 90%
CAS: 6299-02-1 | C8H10ClN | 155.63 g/mol
$262.76 - $262.76
Chemical Identifiers
| CAS | 6299-02-1 |
|---|---|
| Molecular Formula | C8H10ClN |
| Molecular Weight (g/mol) | 155.63 |
| MDL Number | MFCD00044766 |
| InChI Key | PINPOEWMCLFRRB-UHFFFAOYNA-N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC329470050
|
Thermo Scientific Chemicals
329470050 |
5 g |
Each for $262.76
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 6299-02-1 | |
| 100.0 | |
| 1.1000g/mL | |
| Authentic | |
| Glass bottle | |
| 1.5414 | |
| MFCD00044766 | |
| 1.1 | |
| PINPOEWMCLFRRB-UHFFFAOYNA-N | |
| 1-(4-chlorophenyl)ethanamine | |
| 97803 | |
| 90% | |
| 1-(4-Chlorophenyl)ethylamine, 90% |
| 88.0 | |
| Yellow | |
| 232.0°C | |
| 90% | |
| C8H10ClN | |
| ClC6H4CH(CH3)NH2 | |
| 5 g | |
| 1-4-chlorophenyl ethanamine, 1-4-chlorophenyl ethylamine, 4-chloro-alpha-methylbenzylamine, 1-4-chlorophenyl ethan-1-amine, 4-chloro-alpha-methyl benzylamine, 1-4-chlorophenyl-1-ethanamine, 1-4-chloro-phenyl-ethylamine, acmc-20amkz, acmc-20apje, acmc-209jma | |
| CC(C1=CC=C(C=C1)Cl)N | |
| 155.63 | |
| 155.63 | |
| Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 228-576-0
RUO – Research Use Only