missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)ethylamine, 90%

Catalog No. AC329470050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC329470050 5 g
AC329470010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC329470050 Supplier Thermo Scientific Chemicals Supplier No. 329470050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6299-02-1 | C8H10ClN | 155.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6299-02-1
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.63
MDL Number MFCD00044766
InChI Key PINPOEWMCLFRRB-UHFFFAOYNA-N

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Yellow
Density 1.1000g/mL
Boiling Point 232.0°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Refractive Index 1.5414
Linear Formula ClC6H4CH(CH3)NH2
Quantity 5 g
Specific Gravity 1.1
Synonym 1-4-chlorophenyl ethanamine, 1-4-chlorophenyl ethylamine, 4-chloro-alpha-methylbenzylamine, 1-4-chlorophenyl ethan-1-amine, 4-chloro-alpha-methyl benzylamine, 1-4-chlorophenyl-1-ethanamine, 1-4-chloro-phenyl-ethylamine, acmc-20amkz, acmc-20apje, acmc-209jma
SMILES CC(C1=CC=C(C=C1)Cl)N
IUPAC Name 1-(4-chlorophenyl)ethanamine
PubChem CID 97803
Formula Weight 155.63
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 1-(4-Chlorophenyl)ethylamine, 90%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 228-576-0

RUO – Research Use Only