missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Chlorophenyl)-4,4,4-trifluoro-1,3-butanedione 98.0+%, TCI America™
$76.43 - $266.56
Chemical Identifiers
| CAS | 18931-60-7 |
|---|---|
| Molecular Formula | C10H6ClF3O2 |
| Molecular Weight (g/mol) | 250.60 |
| MDL Number | MFCD00511276 |
| InChI Key | RESLPNKXHQNERF-YVMONPNESA-N |
| PubChem CID | 2773832 |
| IUPAC Name | (3Z)-4-(4-chlorophenyl)-1,1,1-trifluoro-4-hydroxybut-3-en-2-one |
| SMILES | O\C(=C/C(=O)C(F)(F)F)C1=CC=C(Cl)C=C1 |
Chemical Identifiers
| 18931-60-7 | |
| 250.60 | |
| RESLPNKXHQNERF-YVMONPNESA-N | |
| (3Z)-4-(4-chlorophenyl)-1,1,1-trifluoro-4-hydroxybut-3-en-2-one |
| C10H6ClF3O2 | |
| MFCD00511276 | |
| 2773832 | |
| O\C(=C/C(=O)C(F)(F)F)C1=CC=C(Cl)C=C1 |
Specifications
| 18931-60-7 | |
| White | |
| MFCD00511276 | |
| RESLPNKXHQNERF-YVMONPNESA-N | |
| (3Z)-4-(4-chlorophenyl)-1,1,1-trifluoro-4-hydroxybut-3-en-2-one | |
| 2773832 | |
| ≥98.0% (GC,T) | |
| 1-(4-Chlorophenyl)-4,4,4-trifluoro-1,3-butanedione |
| 62°C | |
| C10H6ClF3O2 | |
| 1 g | |
| O\C(=C/C(=O)C(F)(F)F)C1=CC=C(Cl)C=C1 | |
| 250.60 | |
| 250.60 | |
| Crystalline Powder |
Safety and Handling
TSCA : No