missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-3-buten-1-ol, 97%

Catalog No. p-7035662
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5338103 1 g
AAH5338106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5338103 Supplier Thermo Scientific Chemicals Supplier No. H5338103

CAS: 14506-33-3 | C10H11ClO | 182.647 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14506-33-3
Molecular Formula C10H11ClO
Molecular Weight (g/mol) 182.647
MDL Number MFCD03427246
InChI Key GJODRJDLQVBTMF-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl-3-buten-1-ol, 1-4-chlorophenyl but-3-en-1-ol, benzenemethanol,4-chloro-a-2-propen-1-yl, 4-4-chlorophenyl-1-buten-4-ol, 1-4-chloro-phenyl-but-3-en-1-ol, acmc-20apb1, 4-chloro-alpha-allylbenzyl alcohol, 4-4-chlorophenyl but-1-en-4-ol, 1-4-chlorophenyl-but-3-en-1-ol
PubChem CID 2757771
IUPAC Name 1-(4-chlorophenyl)but-3-en-1-ol
SMILES C=CCC(C1=CC=C(C=C1)Cl)O

Specifications

Density 1.137
Quantity 1 g
Refractive Index 1.544
Boiling Point 140°C (2 mmHg)
Formula Weight 182.65
Percent Purity 97%
Chemical Name or Material 1-(4-Chlorophenyl)-3-buten-1-ol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only