missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-2,5-dimethylpyrrole 98.0+%, TCI America™

Catalog No. C19515G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C19515G Supplier TCI America Supplier No. C19515G
Only null left

Chemical Identifiers

CAS 5044-23-5
Molecular Formula C12H12ClN
Molecular Weight (g/mol) 205.685
InChI Key NNKFIFOSLJJRES-UHFFFAOYSA-N
Synonym 2,5-Dimethyl-1-(4-chlorophenyl)pyrrole
PubChem CID 78742
IUPAC Name 1-(4-chlorophenyl)-2,5-dimethylpyrrole
SMILES CC1=CC=C(N1C2=CC=C(C=C2)Cl)C

Specifications

Melting Point 47°C
Quantity 5 g
Formula Weight 205.69
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Chlorophenyl)-2,5-dimethylpyrrole