missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Chlorophenyl)-1-cyclopropanecarboxylic Acid 98.0+%, TCI America™
Supplier: TCI America C295425G
Specifications
| 1-(4-Chlorophenyl)-1-cyclopropanecarboxylic Acid | |
| 72934-37-3 | |
| 25 g | |
| C10H9ClO2 | |
| 1-4-chlorophenyl cyclopropanecarboxylic acid, 1-4-chlorophenyl cyclopropane-1-carboxylic acid, 1-4-chlorophenyl-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-4-chlorophenyl, 1-p-chlorophenyl cyclopropanecarboxylic acid, acmc-209oos, 1-4-chlorophenyl cyclopropanecarboxylicacid, 1-4chloro-phenyl-cyclopropylcarboxylic acid, 1-4-chloro-phenyl-cyclopropylcarboxylic acid | |
| C1CC1(C2=CC=C(C=C2)Cl)C(=O)O | |
| 196.63 | |
| 196.63 |
| 154°C | |
| White | |
| Crystal-Powder at 20°C | |
| MFCD00001289 | |
| YAHLWSGIQJATGG-UHFFFAOYSA-N | |
| 1-(4-chlorophenyl)cyclopropane-1-carboxylic acid | |
| 98606 | |
| ≥98.0% (T) |
Chemical Identifiers
| 72934-37-3 | |
| 196.63 | |
| YAHLWSGIQJATGG-UHFFFAOYSA-N | |
| 98606 | |
| C1CC1(C2=CC=C(C=C2)Cl)C(=O)O |
| C10H9ClO2 | |
| MFCD00001289 | |
| 1-4-chlorophenyl cyclopropanecarboxylic acid, 1-4-chlorophenyl cyclopropane-1-carboxylic acid, 1-4-chlorophenyl-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-4-chlorophenyl, 1-p-chlorophenyl cyclopropanecarboxylic acid, acmc-209oos, 1-4-chlorophenyl cyclopropanecarboxylicacid, 1-4chloro-phenyl-cyclopropylcarboxylic acid, 1-4-chloro-phenyl-cyclopropylcarboxylic acid | |
| 1-(4-chlorophenyl)cyclopropane-1-carboxylic acid |
Safety and Handling
TSCA : No