missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-1-cyclopropanecarboxylic Acid 98.0+%, TCI America™

Catalog No. C29545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29545G 5 g
C295425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29545G Supplier TCI America Supplier No. C29545G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 72934-37-3
Molecular Formula C10H9ClO2
Molecular Weight (g/mol) 196.63
MDL Number MFCD00001289
InChI Key YAHLWSGIQJATGG-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl cyclopropanecarboxylic acid, 1-4-chlorophenyl cyclopropane-1-carboxylic acid, 1-4-chlorophenyl-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-4-chlorophenyl, 1-p-chlorophenyl cyclopropanecarboxylic acid, acmc-209oos, 1-4-chlorophenyl cyclopropanecarboxylicacid, 1-4chloro-phenyl-cyclopropylcarboxylic acid, 1-4-chloro-phenyl-cyclopropylcarboxylic acid
PubChem CID 98606
IUPAC Name 1-(4-chlorophenyl)cyclopropane-1-carboxylic acid
SMILES C1CC1(C2=CC=C(C=C2)Cl)C(=O)O

Specifications

Melting Point 154°C
Color White
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 196.63
Percent Purity ≥98.0% (T)
Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclopropanecarboxylic Acid