missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-1-cyclopropanecarboxylic acid, 99%

Catalog No. AC170690500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC170690500 50 g Glass bottle
AC170690100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC170690500 Supplier Thermo Scientific Chemicals Supplier No. 170690500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 72934-37-3 | C10H9ClO2 | 196.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 72934-37-3
Molecular Formula C10H9ClO2
Molecular Weight (g/mol) 196.63
MDL Number MFCD00001289
InChI Key YAHLWSGIQJATGG-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl cyclopropanecarboxylic acid, 1-4-chlorophenyl cyclopropane-1-carboxylic acid, 1-4-chlorophenyl-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-4-chlorophenyl, 1-p-chlorophenyl cyclopropanecarboxylic acid, acmc-209oos, 1-4-chlorophenyl cyclopropanecarboxylicacid, 1-4chloro-phenyl-cyclopropylcarboxylic acid, 1-4-chloro-phenyl-cyclopropylcarboxylic acid
PubChem CID 98606
IUPAC Name 1-(4-chlorophenyl)cyclopropane-1-carboxylic acid
SMILES C1CC1(C2=CC=C(C=C2)Cl)C(=O)O

Specifications

Melting Point 152.0°C to 155.0°C
Color White
CAS Min % 98.5
CAS Max % 100.0
Quantity 50 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 99%
Linear Formula ClC6H4C3H4CO2H
Formula Weight 196.63
Percent Purity 99%
Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclopropanecarboxylic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 277-105-5
TSCA TSCA

RUO – Research Use Only