missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butanesultone 99.0+%, TCI America™

$124.89 - $497.70

Chemical Identifiers

CAS 1633-83-6
Molecular Formula C4H8O3S
Molecular Weight (g/mol) 136.17
MDL Number MFCD00006584
InChI Key MHYFEEDKONKGEB-UHFFFAOYSA-N
Synonym 1,4-butane sultone, 1,4-butanesultone, butane sultone, butanesultone, 1,2-oxathiane, 2,2-dioxide, butanesulfone, 1,2-oxathiane 2,2-dioxide, 1,4-butylene sulfone, delta-valerosultone, 1,4-butanesulfone
PubChem CID 15411
IUPAC Name 1,2λ⁶-oxathiane-2,2-dione
SMILES O=S1(=O)CCCCO1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B013625G
missing translation for 'viewDocuments'
TCI America
B013625G
25 g
Each for $124.89
Only null left
 
B0136250G
missing translation for 'viewDocuments'
TCI America
B0136250G
250 g
Each for $497.70
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1633-83-6
136.17
MHYFEEDKONKGEB-UHFFFAOYSA-N
15411
O=S1(=O)CCCCO1
C4H8O3S
MFCD00006584
1,4-butane sultone, 1,4-butanesultone, butane sultone, butanesultone, 1,2-oxathiane, 2,2-dioxide, butanesulfone, 1,2-oxathiane 2,2-dioxide, 1,4-butylene sulfone, delta-valerosultone, 1,4-butanesulfone
1,2λ⁶-oxathiane-2,2-dione

Specifications

1633-83-6
146°C
MFCD00006584
1,4-butane sultone, 1,4-butanesultone, butane sultone, butanesultone, 1,2-oxathiane, 2,2-dioxide, butanesulfone, 1,2-oxathiane 2,2-dioxide, 1,4-butylene sulfone, delta-valerosultone, 1,4-butanesulfone
O=S1(=O)CCCCO1
136.17
136.17
Liquid
Colorless
C4H8O3S
25 g
MHYFEEDKONKGEB-UHFFFAOYSA-N
1,2λ⁶-oxathiane-2,2-dione
15411
≥99.0% (GC)
1,4-Butanesultone
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0531

RTECSNumber : RP4300000

TSCA : Yes

SDS
Documents

Documents