Learn More
1,4-Butanesultone, 99%
CAS: 1633-83-6 | C4H8O3S | 136.17 g/mol
$96.56 - $247.14
Chemical Identifiers
| CAS | 1633-83-6 |
|---|---|
| Molecular Formula | C4H8O3S |
| Molecular Weight (g/mol) | 136.17 |
| MDL Number | MFCD00006584 |
| InChI Key | MHYFEEDKONKGEB-UHFFFAOYSA-N |
| Synonym | 1,4-butane sultone, 1,4-butanesultone, butane sultone, butanesultone, 1,2-oxathiane, 2,2-dioxide, butanesulfone, 1,2-oxathiane 2,2-dioxide, 1,4-butylene sulfone, delta-valerosultone, 1,4-butanesulfone |
| PubChem CID | 15411 |
| IUPAC Name | oxathiane 2,2-dioxide |
| SMILES | O=S1(=O)CCCCO1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0399214
|
Thermo Scientific Chemicals
L0399214 |
25 g |
Each for $96.56
|
|
|||||
|
AAL0399222
|
Thermo Scientific Chemicals
L0399222 |
100 g |
Each for $247.14
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications1,4-Butanesultone is an alkylating agent used in synthetic chemistry. It serves as a wood preservative. It is employed as an intermediate of the organic sulfonation agent. Further, it is used as a raw material for electroplating intermediate. In addition to this, it finds application in active pharmaceutical ingredients, photographic materials, lithium battery and surface active agents.
Solubility
Miscible with chloroform and methanol. Slightly miscible with water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
| 1633-83-6 | |
| 136.17 | |
| MHYFEEDKONKGEB-UHFFFAOYSA-N | |
| 15411 | |
| O=S1(=O)CCCCO1 |
| C4H8O3S | |
| MFCD00006584 | |
| 1,4-butane sultone, 1,4-butanesultone, butane sultone, butanesultone, 1,2-oxathiane, 2,2-dioxide, butanesulfone, 1,2-oxathiane 2,2-dioxide, 1,4-butylene sulfone, delta-valerosultone, 1,4-butanesulfone | |
| oxathiane 2,2-dioxide |
Specifications
| 1633-83-6 | |
| 1.335 | |
| 153°C (307°F) | |
| 1.464 | |
| 25 g | |
| Moisture sensitive | |
| Miscible with chloroform and methanol. Slightly miscible with water. | |
| O=S1(=O)CCCCO1 | |
| 136.17 | |
| 136.17 | |
| 1,4-Butanesultone |
| 12°C to 15°C | |
| 114°C to 116°C (1.5 mmHg) | |
| C4H8O3S | |
| MFCD00006584 | |
| 110588 | |
| 1,4-butane sultone, 1,4-butanesultone, butane sultone, butanesultone, 1,2-oxathiane, 2,2-dioxide, butanesulfone, 1,2-oxathiane 2,2-dioxide, 1,4-butylene sulfone, delta-valerosultone, 1,4-butanesulfone | |
| MHYFEEDKONKGEB-UHFFFAOYSA-N | |
| oxathiane 2,2-dioxide | |
| 15411 | |
| 99% |
Safety and Handling
GHS H Statement
H341-H302
Suspected of causing genetic defects.
Harmful if swallowed.
P201-P202-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
H302-H315-H320-H335-H341-H351
EINECSNumber : 216-647-9
RTECSNumber : RP4300000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only