missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Butanesultone, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
1,4-Butanesultone is an alkylating agent used in synthetic chemistry. It serves as a wood preservative. It is employed as an intermediate of the organic sulfonation agent. Further, it is used as a raw material for electroplating intermediate. In addition to this, it finds application in active pharmaceutical ingredients, photographic materials, lithium battery and surface active agents.
Solubility
Miscible with chloroform and methanol. Slightly miscible with water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
1,4-Butanesultone is an alkylating agent used in synthetic chemistry. It serves as a wood preservative. It is employed as an intermediate of the organic sulfonation agent. Further, it is used as a raw material for electroplating intermediate. In addition to this, it finds application in active pharmaceutical ingredients, photographic materials, lithium battery and surface active agents.
Solubility
Miscible with chloroform and methanol. Slightly miscible with water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
Specifications
Specifications
| Melting Point | 12°C to 15°C |
| Density | 1.335 |
| Boiling Point | 114°C to 116°C (1.5 mmHg) |
| Flash Point | 153°C (307°F) |
| Refractive Index | 1.464 |
| Quantity | 25 g |
| Beilstein | 110588 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Miscible with chloroform and methanol. Slightly miscible with water. |
| Formula Weight | 136.17 |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | H302-H315-H320-H335-H341-H351 |
| missing translation for 'healthHazard2' | GHS H Statement H341-H302 Suspected of causing genetic defects. Harmful if swallowed. |
| missing translation for 'healthHazard3' | P201-P202-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 216-647-9 |
| RTECSNumber | RP4300000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only