missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Butanedithiol 95.0+%, TCI America™
Supplier: TCI America B08295ML
Specifications
| 1,4-Butanedithiol | |
| Yellow | |
| C4H10S2 | |
| 5 mL | |
| SMTOKHQOVJRXLK-UHFFFAOYSA-N | |
| butane-1,4-dithiol | |
| 79148 | |
| ≥95.0% (GC,T) |
| 1191-08-8 | |
| 195°C | |
| MFCD00004906 | |
| 1,4-butanedithiol, 1,4-dimercaptobutane, tetramethylenedithiol, unii-7jd227iumc, tetramethylene dimercaptan, 7jd227iumc, 1,4-butane dithiol, 1,4-dibutyl mercaptan, acmc-1bs9f | |
| SCCCCS | |
| 122.24 | |
| 122.24 | |
| Liquid |
Chemical Identifiers
| 1191-08-8 | |
| 122.24 | |
| SMTOKHQOVJRXLK-UHFFFAOYSA-N | |
| 79148 | |
| SCCCCS |
| C4H10S2 | |
| MFCD00004906 | |
| 1,4-butanedithiol, 1,4-dimercaptobutane, tetramethylenedithiol, unii-7jd227iumc, tetramethylene dimercaptan, 7jd227iumc, 1,4-butane dithiol, 1,4-dibutyl mercaptan, acmc-1bs9f | |
| butane-1,4-dithiol |
Safety and Handling
EINECSNumber : (2)-2448
TSCA : Yes