missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butanediol Diglycidyl Ether 93.0+%, TCI America™

Catalog No. p-7118769
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
250 mL
Catalog No. Quantity
B096425ML 25 mL
B0964250ML 250 mL
2 options

Catalog No. B096425ML

Supplier: TCI America B096425ML

Only null left

Chemical Identifiers

CAS 2425-79-8
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00005146
InChI Key SHKUUQIDMUMQQK-UHFFFAOYNA-N
Synonym 1,4-butanediol diglycidyl ether, 1,4-diglycidyloxybutane, 1,4-bis glycidyloxy butane, butanediol diglycidyl ether, araldit dy 026, 1,4-butane diglycidyl ether, grilonit rv 1806, 1,4-diglycidloxybutane, chs-rr2, 1,4-bis 2,3-epoxypropoxy butane
PubChem CID 17046
IUPAC Name 2-({4-[(oxiran-2-yl)methoxy]butoxy}methyl)oxirane
SMILES C(CCOCC1CO1)COCC1CO1

Specifications

Melting Point -21°C
Color Yellow
Boiling Point 160°C
Quantity 25 mL
Formula Weight 202.25
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Butanediol Diglycidyl Ether
EINECSNumber (2)-0396
RTECSNumber EJ5100000
TSCA Yes