missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 1,4-Butanediol diglycidyl ether

Catalog No. 220892250G
Encompass
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
220892250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 220892250G Supplier Sigma Aldrich Supplier No. 220892250G
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,4-Butanediol diglycidyl ether (BDDE) is a viscous liquid , which is hygroscopic in nature. BDDE is the most commonly used homobifunctional epoxide compound. BDDE may react with amines, hydroxyls, sulfhydryl groups to produce secondary amines, ether or thioether bonds respectively.BDDE is a cross linking agent used for preparing amylose, xylan and hydroxyethyl-celluloseto cross link polyethylenimine which also serves as a hole-blocking layer in organic solar cellsto cross link hyaluronic acid into hydrogelsfor the activation of soluble dextran polymers.Amine or hydroxyl containing bis-epoxy supports are reacted with BDDE to form particles containing terminal epoxide group for immobilization reactions.

Specifications

CAS 2425-79-8
Density 1.1 g/mL (at 25°C)
Boiling Point 266°C
Molecular Formula C10H18O4
Refractive Index n20/D 1.453 (literature)
Linear Formula C10H18O4
Quantity 250 g
MDL Number MFCD00005146
Synonym 1,4-Bis(2,3-epoxypropyloxy)butane
Molecular Weight (g/mol) 202.25
Percent Purity >=95%
Show More Show Less