missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butanediol Bis(thioglycolate) 95.0+%, TCI America™

Catalog No. B3423500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3423500G 500 g
B342325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3423500G Supplier TCI America Supplier No. B3423500G
Only null left

Chemical Identifiers

CAS 10193-95-0
Molecular Formula C8H14O4S2
Molecular Weight (g/mol) 238.32
MDL Number MFCD00266655
InChI Key IPNDIMIIGZSERC-UHFFFAOYSA-N
Synonym 1,4-Bis(mercaptoacetoxy)butane
PubChem CID 25040
IUPAC Name 4-[(2-sulfanylacetyl)oxy]butyl 2-sulfanylacetate
SMILES SCC(=O)OCCCCOC(=O)CS

Specifications

Color Yellow
Quantity 500 g
Formula Weight 238.32
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1,4-Butanediol Bis(thioglycolate)
EINECSNumber (2)-1360
RTECSNumber AI8300000
TSCA Yes