missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butanediol Bis(3-aminopropyl) Ether 98.0+%, TCI America™

Catalog No. B1201500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1201500ML 500 mL
B120125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1201500ML Supplier TCI America Supplier No. B1201500ML

Chemical Identifiers

CAS 7300-34-7
Molecular Formula C10H24N2O2
Molecular Weight (g/mol) 204.31
MDL Number MFCD00009802
InChI Key YOOSAIJKYCBPFW-UHFFFAOYSA-N
Synonym 1,4-bis 3-aminopropoxy butane, 4,9-dioxa-1,12-dodecanediamine, 4,9-dioxadodecane-1,12-diamine, 1,12-diamino-4,9-dioxadodecane, 1,4-butanediol bis 3-aminopropyl ether, 3,3'-tetramethylenedioxy bis propylamine, 1,4-bis gamma-aminopropoxy butane, alpha,omega-diamino-4,9-dioxadodecane, 3,3'-butane-1,4-diylbis oxy bis propan-1-amine, 1,4-butanediol bis gamma-aminopropyl ether
PubChem CID 81729
IUPAC Name 3-[4-(3-aminopropoxy)butoxy]propan-1-amine
SMILES NCCCOCCCCOCCCN

Specifications

Color Yellow
Boiling Point 136°C
Quantity 500 mL
UN Number 2735
Formula Weight 204.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Butanediol Bis(3-aminopropyl) Ether
EINECSNumber (2)-2435&(7)-0166
RTECSNumber TX1515087
TSCA Yes