missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butanediol Bis(3-aminocrotonate) 96.0+%, TCI America™

Catalog No. B1069500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1069500G 500 g
B106925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1069500G Supplier TCI America Supplier No. B1069500G
Only null left

Chemical Identifiers

CAS 14205-47-1
Molecular Formula C12H20N2O4
Molecular Weight (g/mol) 256.302
MDL Number MFCD00059862
InChI Key DFUXQARZEWMFLQ-FKJILZIQSA-N
Synonym 1,4-Butylene Bis(3-aminocrotonate)
PubChem CID 22186498
IUPAC Name 4-[(E)-3-aminobut-2-enoyl]oxybutyl (Z)-3-aminobut-2-enoate
SMILES CC(=CC(=O)OCCCCOC(=O)C=C(C)N)N

Specifications

Melting Point 148°C
Color Yellow
Quantity 500 g
Formula Weight 256.30
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Butanediol Bis(3-aminocrotonate)