Learn More
1,4-Butane sultone, 99+%
CAS: 1633-83-6 | C4H8O3S | 136.17 g/mol
Supplier: Thermo Scientific Chemicals 107671000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1, 4-Butane sultone | |
| 1633-83-6 | |
| 100.0 | |
| Colorless to Yellow | |
| 153°C (19.0 HPA) | |
| Authentic | |
| Glass bottle | |
| 1.4600 to 1.4640 | |
| 100 mL | |
| 1.331 | |
| Solubility in water: 54g/L (20°C), decomposes. Other solubilities: soluble in chloroform | |
| O=S1(=O)CCCCO1 | |
| 136.17 | |
| 136.17 | |
| Liquid |
| 99+% | |
| 99.0 | |
| 12.5°C to 15.0°C | |
| 1.3310g/mL | |
| 154°C | |
| 99% min. (GC) | |
| C4H8O3S | |
| MFCD00006584 | |
| 19, V,1, 10 | |
| 1,4-butane sultone, 1,4-butanesultone, butane sultone, butanesultone, 1,2-oxathiane, 2,2-dioxide, butanesulfone, 1,2-oxathiane 2,2-dioxide, 1,4-butylene sulfone, delta-valerosultone, 1,4-butanesulfone | |
| MHYFEEDKONKGEB-UHFFFAOYSA-N | |
| oxathiane 2,2-dioxide | |
| 15411 | |
| 99+% |
Chemical Identifiers
| 1633-83-6 | |
| 136.17 | |
| MHYFEEDKONKGEB-UHFFFAOYSA-N | |
| 15411 | |
| O=S1(=O)CCCCO1 |
| C4H8O3S | |
| MFCD00006584 | |
| 1,4-butane sultone, 1,4-butanesultone, butane sultone, butanesultone, 1,2-oxathiane, 2,2-dioxide, butanesulfone, 1,2-oxathiane 2,2-dioxide, 1,4-butylene sulfone, delta-valerosultone, 1,4-butanesulfone | |
| oxathiane 2,2-dioxide |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Suspected of causing genetic defects if inhaled.
GHS P Statement
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 216-647-9
RTECSNumber : RP4300000
TSCA : TSCA
RUO – Research Use Only