missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromophenyl)piperidine 98.0+%, TCI America™

Catalog No. B39305G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39305G 5 g
B39301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39305G Supplier TCI America Supplier No. B39305G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22148-20-5
Molecular Formula C11H14BrN
Molecular Weight (g/mol) 240.144
MDL Number MFCD04112533
InChI Key OACWQRGNHICZFD-UHFFFAOYSA-N
Synonym 1-4-bromophenyl piperidine, 1-4-bromo-phenyl-piperidine, piperidine, 1-4-bromophenyl, piperidine,1-4-bromophenyl, acmc-1cpan, 4-bromophenyl piperidine, n-4-bromophenyl piperidine, 1-piperidino-4-bromobenzene, amth037
PubChem CID 7016460
IUPAC Name 1-(4-bromophenyl)piperidine
SMILES C1CCN(CC1)C2=CC=C(C=C2)Br

Specifications

Melting Point 76°C
Color White
Quantity 5 g
Formula Weight 240.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Bromophenyl)piperidine