missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromophenyl)piperidine, 97%

Catalog No. p-7038193
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAH5710203 1 g
AAH5710206 5 g
2 options

Catalog No. AAH5710203

Supplier: Thermo Scientific Chemicals H5710203

Only null left

CAS: 22148-20-5 | C11H14BrN | 240.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22148-20-5
Molecular Formula C11H14BrN
Molecular Weight (g/mol) 240.144
MDL Number MFCD04112533
InChI Key OACWQRGNHICZFD-UHFFFAOYSA-N
Synonym 1-4-bromophenyl piperidine, 1-4-bromo-phenyl-piperidine, piperidine, 1-4-bromophenyl, piperidine,1-4-bromophenyl, acmc-1cpan, 4-bromophenyl piperidine, n-4-bromophenyl piperidine, 1-piperidino-4-bromobenzene, amth037
PubChem CID 7016460
IUPAC Name 1-(4-bromophenyl)piperidine
SMILES C1CCN(CC1)C2=CC=C(C=C2)Br

Specifications

Quantity 1 g
Formula Weight 240.14
Percent Purity 97%
Chemical Name or Material 1-(4-Bromophenyl)piperidine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only