missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromophenyl)octane, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB09864EE

Quantity 50g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. BTB09864EE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

51554-93-9
269.226
OOZQSVXPBCINJF-UHFFFAOYSA-N
142854
CCCCCCCCC1=CC=C(C=C1)Br
C14H21Br
MFCD00038348
1-4-bromophenyl octane, 1-bromo-4-n-octylbenzene, p-octylbromobenzene, p-bromophenyloctane, 4-n-octylbromobenzene, 1-p-bromophenyl octane, 4-bromooctylbenzene, 4-oct-1-yl bromobenzene, benzene, 1-bromo-4-octyl, 4-bromo-1-octylbenzene
1-bromo-4-octylbenzene

Specifications

1-(4-Bromophenyl)octane
96.5
97%
C14H21Br
50g
1-4-bromophenyl octane, 1-bromo-4-n-octylbenzene, p-octylbromobenzene, p-bromophenyloctane, 4-n-octylbromobenzene, 1-p-bromophenyl octane, 4-bromooctylbenzene, 4-oct-1-yl bromobenzene, benzene, 1-bromo-4-octyl, 4-bromo-1-octylbenzene
CCCCCCCCC1=CC=C(C=C1)Br
269.226
269.22
51554-93-9
100.0
Amber Glass Bottle
BrC6H4(CH2)7CH3
MFCD00038348
OOZQSVXPBCINJF-UHFFFAOYSA-N
1-bromo-4-octylbenzene
142854
97%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents