missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromophenyl)-2-phenylbenzimidazole 98.0+%, TCI America™

Catalog No. B49295G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49295G 5 g
B49291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49295G Supplier TCI America Supplier No. B49295G
Only null left

Chemical Identifiers

CAS 760212-58-6
Molecular Formula C19H13BrN2
Molecular Weight (g/mol) 349.23
MDL Number MFCD09746342
InChI Key PPYIZNYOMNYZCG-UHFFFAOYSA-N
PubChem CID 23094071
IUPAC Name 1-(4-bromophenyl)-2-phenyl-1H-1,3-benzodiazole
SMILES BrC1=CC=C(C=C1)N1C2=CC=CC=C2N=C1C1=CC=CC=C1

Specifications

Melting Point 137°C
Quantity 5 g
Formula Weight 349.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Bromophenyl)-2-phenylbenzimidazole