missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromophenoxy)hexane, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB09795EE

Quantity 50g

 View more versions of this product

Catalog No. BTB09795EE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

30752-19-3
257.171
1-bromo-4-hexyloxy benzene, 1-bromo-4-hexyloxybenzene, 1-4-bromophenoxy hexane, 1-bromo-4-n-hexyloxybenzene, 4-n-hexyloxybromobenzene, p-bromophenylhexyl ether, benzene, 1-bromo-4-hexyloxy, p-hexyloxybromobenzene, 4-hexyloxybromobenzene, 4-bromophenyl hexyl ether
1-bromo-4-hexoxybenzene
C12H17BrO
GKLMJONYGGTHHM-UHFFFAOYSA-N
520492
CCCCCCOC1=CC=C(C=C1)Br

Specifications

1-(4-Bromophenoxy)hexane
97%
50g
GKLMJONYGGTHHM-UHFFFAOYSA-N
1-bromo-4-hexoxybenzene
520492
30752-19-3
C12H17BrO
1-bromo-4-hexyloxy benzene, 1-bromo-4-hexyloxybenzene, 1-4-bromophenoxy hexane, 1-bromo-4-n-hexyloxybenzene, 4-n-hexyloxybromobenzene, p-bromophenylhexyl ether, benzene, 1-bromo-4-hexyloxy, p-hexyloxybromobenzene, 4-hexyloxybromobenzene, 4-bromophenyl hexyl ether
CCCCCCOC1=CC=C(C=C1)Br
257.171
257.17
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents