missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Bis(trimethylsilyl)-1,3-butadiyne 99.0+%, TCI America™
$171.49 - $548.03
Chemical Identifiers
| CAS | 4526-07-2 |
|---|---|
| Molecular Formula | C10H18Si2 |
| Molecular Weight (g/mol) | 194.42 |
| MDL Number | MFCD00009839 |
| InChI Key | LBNVCJHJRYJVPK-UHFFFAOYSA-N |
| Synonym | 1,4-bis trimethylsilyl-1,3-butadiyne, 1,4-bis trimethylsilyl butadiyne, 1,4-bis trimethylsilyl buta-1,3-diyne, trimethyl 4-trimethylsilylbuta-1,3-diynyl silane, btmsbd, acmc-1ctvu, bis trimethylsiyl butadiyne, ksc491i6r, bis trimethylsilyl-1,3-butadiyne, bis trimethylsilyl butadiyne |
| PubChem CID | 138279 |
| IUPAC Name | trimethyl(4-trimethylsilylbuta-1,3-diynyl)silane |
| SMILES | C[Si](C)(C)C#CC#C[Si](C)(C)C |
Chemical Identifiers
| 4526-07-2 | |
| 194.42 | |
| LBNVCJHJRYJVPK-UHFFFAOYSA-N | |
| 138279 | |
| C[Si](C)(C)C#CC#C[Si](C)(C)C |
| C10H18Si2 | |
| MFCD00009839 | |
| 1,4-bis trimethylsilyl-1,3-butadiyne, 1,4-bis trimethylsilyl butadiyne, 1,4-bis trimethylsilyl buta-1,3-diyne, trimethyl 4-trimethylsilylbuta-1,3-diynyl silane, btmsbd, acmc-1ctvu, bis trimethylsiyl butadiyne, ksc491i6r, bis trimethylsilyl-1,3-butadiyne, bis trimethylsilyl butadiyne | |
| trimethyl(4-trimethylsilylbuta-1,3-diynyl)silane |
Specifications
| 4526-07-2 | |
| C10H18Si2 | |
| 1 g | |
| LBNVCJHJRYJVPK-UHFFFAOYSA-N | |
| trimethyl(4-trimethylsilylbuta-1,3-diynyl)silane | |
| 138279 | |
| ≥99.0% (GC) | |
| 1,4-Bis(trimethylsilyl)-1,3-butadiyne |
| 112°C | |
| MFCD00009839 | |
| 1,4-bis trimethylsilyl-1,3-butadiyne, 1,4-bis trimethylsilyl butadiyne, 1,4-bis trimethylsilyl buta-1,3-diyne, trimethyl 4-trimethylsilylbuta-1,3-diynyl silane, btmsbd, acmc-1ctvu, bis trimethylsiyl butadiyne, ksc491i6r, bis trimethylsilyl-1,3-butadiyne, bis trimethylsilyl butadiyne | |
| C[Si](C)(C)C#CC#C[Si](C)(C)C | |
| 194.42 | |
| 194.42 | |
| Crystalline Powder |
Safety and Handling
TSCA : No