missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(trimethylsilyl)-1,3-butadiyne 99.0+%, TCI America™

Catalog No. B12985G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12985G 5 g
B12981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12985G Supplier TCI America Supplier No. B12985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4526-07-2
Molecular Formula C10H18Si2
Molecular Weight (g/mol) 194.42
MDL Number MFCD00009839
InChI Key LBNVCJHJRYJVPK-UHFFFAOYSA-N
Synonym 1,4-bis trimethylsilyl-1,3-butadiyne, 1,4-bis trimethylsilyl butadiyne, 1,4-bis trimethylsilyl buta-1,3-diyne, trimethyl 4-trimethylsilylbuta-1,3-diynyl silane, btmsbd, acmc-1ctvu, bis trimethylsiyl butadiyne, ksc491i6r, bis trimethylsilyl-1,3-butadiyne, bis trimethylsilyl butadiyne
PubChem CID 138279
IUPAC Name trimethyl(4-trimethylsilylbuta-1,3-diynyl)silane
SMILES C[Si](C)(C)C#CC#C[Si](C)(C)C

Specifications

Melting Point 112°C
Quantity 5 g
Formula Weight 194.42
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis(trimethylsilyl)-1,3-butadiyne