missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(maleimido)butane 98.0+%, TCI America™

Catalog No. p-7123177
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3805100MG 100 mg
B38051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3805100MG Supplier TCI America Supplier No. B3805100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 28537-70-4
Molecular Formula C12H12N2O4
Molecular Weight (g/mol) 248.238
MDL Number MFCD01318342
InChI Key WXXSHAKLDCERGU-UHFFFAOYSA-N
Synonym N,N′C-Tetramethylenebismaleimide, N,N′C-Tetramethylenedimaleimide, BMB
PubChem CID 239576
IUPAC Name 1-[4-(2,5-dioxopyrrol-1-yl)butyl]pyrrole-2,5-dione
SMILES C1=CC(=O)N(C1=O)CCCCN2C(=O)C=CC2=O

Specifications

Melting Point 206°C
Quantity 100 mg
Formula Weight 248.24
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis(maleimido)butane
TSCA No
Recommended Storage Freezer