missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(4-Chlorobenzhydryl)Piperazine, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022896
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022896 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022896 Supplier Merck Emd Millipore Supplier No. BP441

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

CAS 346451-15-8
Molecular Formula C30H28Cl2N2
Linear Formula C30H28Cl2N2
Quantity 25 mg
Formula Weight 487.46
Grade BP