missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(3-aminopropyl)piperazine 98.0+%, TCI America™

Catalog No. B1041500ML
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1041500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1041500ML Supplier TCI America Supplier No. B1041500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7209-38-3
Molecular Formula C10H24N4
Molecular Weight (g/mol) 200.33
MDL Number MFCD00006161
InChI Key XUSNPFGLKGCWGN-UHFFFAOYSA-N
Synonym 1,4-bis 3-aminopropyl piperazine, 1,4-piperazinedipropanamine, 3,3'-piperazine-1,4-diyl bis propan-1-amine, bis aminopropyl piperazine, n,n'-bis 3-aminopropyl piperazine, unii-7gvx6c734b, bapp, ccris 8915, 1,4-di 3-aminopropyl piperazine, 3,3'-piperazine-1,4-diyldipropan-1-amine
PubChem CID 81629
IUPAC Name 3-[4-(3-aminopropyl)piperazin-1-yl]propan-1-amine
SMILES C1CN(CCN1CCCN)CCCN

Specifications

Color Yellow
Boiling Point 158°C
Quantity 500 mL
UN Number 3267
Formula Weight 200.33
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1,4-Bis(3-aminopropyl)piperazine
EINECSNumber (5)-0962
TSCA Yes