Learn More
1,4-Bis(3-aminopropyl)piperazine, 98%
CAS: 7209-38-3 | C10H24N4 | 200.33 g/mol
$259.49 - $259.49
Chemical Identifiers
| CAS | 7209-38-3 |
|---|---|
| Molecular Formula | C10H24N4 |
| Molecular Weight (g/mol) | 200.33 |
| MDL Number | MFCD00006161 |
| InChI Key | XUSNPFGLKGCWGN-UHFFFAOYSA-N |
| Synonym | 1,4-bis 3-aminopropyl piperazine, 1,4-piperazinedipropanamine, 3,3'-piperazine-1,4-diyl bis propan-1-amine, bis aminopropyl piperazine, n,n'-bis 3-aminopropyl piperazine, unii-7gvx6c734b, bapp, ccris 8915, 1,4-di 3-aminopropyl piperazine, 3,3'-piperazine-1,4-diyldipropan-1-amine |
| PubChem CID | 81629 |
| IUPAC Name | 3-[4-(3-aminopropyl)piperazin-1-yl]propan-1-amine |
| SMILES | C1CN(CCN1CCCN)CCCN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1250822
|
Thermo Scientific Chemicals
L1250822 |
100 g |
Each for $259.49
|
|
|||||
Description
1,4-Bis(3-aminopropyl)piperazine was used to functionalize natural montmorillonite (M) and synthetic kanemite (K)1.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,4-Bis(3-aminopropyl)piperazine was used to functionalize natural montmorillonite (M) and synthetic kanemite (K)1.
Solubility
Fully miscible in water.
Notes
Air Sensitive. Incompatible with oxidizing agents.
Chemical Identifiers
| 7209-38-3 | |
| 200.33 | |
| XUSNPFGLKGCWGN-UHFFFAOYSA-N | |
| 81629 | |
| C1CN(CCN1CCCN)CCCN |
| C10H24N4 | |
| MFCD00006161 | |
| 1,4-bis 3-aminopropyl piperazine, 1,4-piperazinedipropanamine, 3,3'-piperazine-1,4-diyl bis propan-1-amine, bis aminopropyl piperazine, n,n'-bis 3-aminopropyl piperazine, unii-7gvx6c734b, bapp, ccris 8915, 1,4-di 3-aminopropyl piperazine, 3,3'-piperazine-1,4-diyldipropan-1-amine | |
| 3-[4-(3-aminopropyl)piperazin-1-yl]propan-1-amine |
Specifications
| 7209-38-3 | |
| 0.973 | |
| 162°C (323°F) | |
| 1.5018 | |
| 100 g | |
| 111747 | |
| 1,4-bis 3-aminopropyl piperazine, 1,4-piperazinedipropanamine, 3,3'-piperazine-1,4-diyl bis propan-1-amine, bis aminopropyl piperazine, n,n'-bis 3-aminopropyl piperazine, unii-7gvx6c734b, bapp, ccris 8915, 1,4-di 3-aminopropyl piperazine, 3,3'-piperazine-1,4-diyldipropan-1-amine | |
| XUSNPFGLKGCWGN-UHFFFAOYSA-N | |
| 3-[4-(3-aminopropyl)piperazin-1-yl]propan-1-amine | |
| 81629 | |
| 98% |
| 14°C | |
| 150°C to 152°C (2 mmHg) | |
| C10H24N4 | |
| MFCD00006161 | |
| UN2735 | |
| Air Sensitive | |
| Fully miscible in water. | |
| C1CN(CCN1CCCN)CCCN | |
| 200.33 | |
| 200.33 | |
| 1,4-Bis(3-aminopropyl)piperazine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 230-589-1
RTECSNumber : TK8700000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only