missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Bis(2-methylstyryl)benzene 99.0+%, TCI America™
$91.33 - $367.22
Chemical Identifiers
| CAS | 13280-61-0 |
|---|---|
| Molecular Formula | C24H22 |
| Molecular Weight (g/mol) | 310.44 |
| MDL Number | MFCD00008529 |
| InChI Key | QKLPIYTUUFFRLV-YTEMWHBBSA-N |
| Synonym | 1,4-bis 2-methylstyryl benzene, bis-msb, p-bis o-methylstyryl benzene, benzene, 1,4-bis 2-2-methylphenyl ethenyl, 1,4-di 2-methylstyryl benzene, 1,4-bis 2-methylstyryl benzene bis-msb, benzene, p-bis o-methylstyryl, 1,4-di-2-methylstyryl benzene, 2-menthylstyrene, 1-methyl-2-2-4-2-2-methylphenyl ethenyl phenyl ethenyl benzene |
| PubChem CID | 5378735 |
| IUPAC Name | 1,4-bis[(E)-2-(2-methylphenyl)ethenyl]benzene |
| SMILES | CC1=CC=CC=C1C=CC2=CC=C(C=C2)C=CC3=CC=CC=C3C |
Chemical Identifiers
| 13280-61-0 | |
| 310.44 | |
| QKLPIYTUUFFRLV-YTEMWHBBSA-N | |
| 5378735 | |
| CC1=CC=CC=C1C=CC2=CC=C(C=C2)C=CC3=CC=CC=C3C |
| C24H22 | |
| MFCD00008529 | |
| 1,4-bis 2-methylstyryl benzene, bis-msb, p-bis o-methylstyryl benzene, benzene, 1,4-bis 2-2-methylphenyl ethenyl, 1,4-di 2-methylstyryl benzene, 1,4-bis 2-methylstyryl benzene bis-msb, benzene, p-bis o-methylstyryl, 1,4-di-2-methylstyryl benzene, 2-menthylstyrene, 1-methyl-2-2-4-2-2-methylphenyl ethenyl phenyl ethenyl benzene | |
| 1,4-bis[(E)-2-(2-methylphenyl)ethenyl]benzene |
Specifications
| 13280-61-0 | |
| C24H22 | |
| 5 g | |
| QKLPIYTUUFFRLV-YTEMWHBBSA-N | |
| 1,4-bis[(E)-2-(2-methylphenyl)ethenyl]benzene | |
| 5378735 | |
| ≥99.0% (GC) | |
| 1,4-Bis(2-methylstyryl)benzene [Solute for Liquid Scintillation Counting] |
| 179°C | |
| MFCD00008529 | |
| 1,4-bis 2-methylstyryl benzene, bis-msb, p-bis o-methylstyryl benzene, benzene, 1,4-bis 2-2-methylphenyl ethenyl, 1,4-di 2-methylstyryl benzene, 1,4-bis 2-methylstyryl benzene bis-msb, benzene, p-bis o-methylstyryl, 1,4-di-2-methylstyryl benzene, 2-menthylstyrene, 1-methyl-2-2-4-2-2-methylphenyl ethenyl phenyl ethenyl benzene | |
| CC1=CC=CC=C1C=CC2=CC=C(C=C2)C=CC3=CC=CC=C3C | |
| 310.44 | |
| 310.44 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes