missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(2-hydroxyethyl)piperazine 98.0+%, TCI America™

Catalog No. B49035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49035G 5 g
B490325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49035G Supplier TCI America Supplier No. B49035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 122-96-3
Molecular Formula C8H18N2O2
Molecular Weight (g/mol) 174.244
MDL Number MFCD00006157
InChI Key VARKIGWTYBUWNT-UHFFFAOYSA-N
Synonym 1,4-piperazinediethanol, 2,2'-piperazine-1,4-diyl diethanol, 1,4-bis 2-hydroxyethyl piperazine, n,n'-bis 2-hydroxyethyl piperazine, 1,4-di 2-hydroxyethyl piperazine, n,n'-dihydroxyethylpiperazine, n,n'-di 2-hydroxyethyl piperazine, unii-eq349pkx8y, piperazine, n,n'-bis 2-hydroxyethyl, 1,4-bis beta-hydroxyethyl piperazine
PubChem CID 67151
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanol
SMILES C1CN(CCN1CCO)CCO

Specifications

Melting Point 138°C
Color White
Boiling Point 278°C
Quantity 5 g
Formula Weight 174.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis(2-hydroxyethyl)piperazine
EINECSNumber (7)-1365
RTECSNumber TL3675000
TSCA Yes