missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis[2-(5-phenyloxazolyl)]benzene 98.0+%, TCI America™

Catalog No. p-7118488
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B05091G 1 g
B05095G 5 g
B050925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B05091G Supplier TCI America Supplier No. B05091G
Only null left

Chemical Identifiers

CAS 1806-34-4
Molecular Formula C24H16N2O2
Molecular Weight (g/mol) 364.404
MDL Number MFCD00005309
InChI Key MASVCBBIUQRUKL-UHFFFAOYSA-N
Synonym popop, 1,4-bis 5-phenyloxazol-2-yl benzene, oxazole, 2,2'-1,4-phenylene bis 5-phenyl, p-bis 5-phenyloxazol-2-yl benzene, 2,2'-p-phenylenebis 5-phenyloxazole, 1,4-bis 5-phenyl-2-oxazolyl benzene, 5-phenyl-2-4-5-phenyl-1,3-oxazol-2-yl phenyl-1,3-oxazole, 1,4-di 5-phenyl-2-oxazolyl benzene, 1,4-bis 2-5-phenyloxazolyl benzene, oxazole, 2,2'-p-phenylenebis 5-phenyl
PubChem CID 15732
ChEBI CHEBI:52236
IUPAC Name 5-phenyl-2-[4-(5-phenyl-1,3-oxazol-2-yl)phenyl]-1,3-oxazole
SMILES C1=CC=C(C=C1)C2=CN=C(O2)C3=CC=C(C=C3)C4=NC=C(O4)C5=CC=CC=C5

Specifications

Melting Point 245°C
Quantity 1 g
Formula Weight 364.40
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis[2-(5-phenyloxazolyl)]benzene [for scintillation spectrometry]