missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis[(1H-imidazol-1-yl)methyl]benzene 98.0+%, TCI America™

Catalog No. p-7123306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40231G 1 g
B40235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40231G Supplier TCI America Supplier No. B40231G
Only null left

Chemical Identifiers

CAS 56643-83-5
Molecular Formula C14H14N4
Molecular Weight (g/mol) 238.29
MDL Number MFCD06598031
InChI Key NKUFFYFOBGGDTP-UHFFFAOYSA-N
PubChem CID 1257389
IUPAC Name 1-({4-[(1H-imidazol-1-yl)methyl]phenyl}methyl)-1H-imidazole
SMILES C(N1C=CN=C1)C1=CC=C(CN2C=CN=C2)C=C1

Specifications

Melting Point 132°C
Color White
Quantity 1 g
Synonym alpha,alpha′C-Bis[(1-imidazolyl)methyl]-p-xylene
Formula Weight 238.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis[(1H-imidazol-1-yl)methyl]benzene