missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzoquinone, 98+%

1, 4-Benzoquinone, 99%, C6H4O2, CAS Number-106-51-4, 1,4-benzoquine, 1,4-benzoquinone, 2,5-cyclohexadiene-1,4-dione, benzoquinone, chinone, cyclohexadienedione, para-quinone, p-benzoquinone, p-quinone, quinone, 1kg, 98.5% min. (Iodometry), 07,609, 100.0, CHEBI:16509, Yellow to Green, 1.3100g/mL | CAS: 106-51-4 | C6H4O2 | 108.1 g/mol

Supplier:  Thermo Scientific Chemicals 105630250

 View more versions of this product

Catalog No. AC105630250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1, 4-Benzoquinone
98.5
112°C to 116°C
1.3100g/mL
98.5% min. (Iodometry)
C6H4O2
MFCD00001591
07,609
1.31
p-benzoquinone, benzoquinone, quinone, 1,4-benzoquinone, p-quinone, chinone, 2,5-cyclohexadiene-1,4-dione, cyclohexadienedione, para-quinone, 1,4-benzoquine
AZQWKYJCGOJGHM-UHFFFAOYSA-N
cyclohexa-2,5-diene-1,4-dione
4650
108.1
Powder
106-51-4
100.0
Yellow to Green
Authentic
Fibre drum
C6H4(=O)2
25 kg
01,49; 07,55
15,8191
Solubility in water: 10g/L water (25°C). Other solubilities: soluble in alkalies,soluble in alcohol and ether
C1=CC(=O)C=CC1=O
108.1
CHEBI:16509
99%

Chemical Identifiers

106-51-4
108.1
AZQWKYJCGOJGHM-UHFFFAOYSA-N
4650
cyclohexa-2,5-diene-1,4-dione
C6H4O2
MFCD00001591
p-benzoquinone, benzoquinone, quinone, 1,4-benzoquinone, p-quinone, chinone, 2,5-cyclohexadiene-1,4-dione, cyclohexadienedione, para-quinone, 1,4-benzoquine
CHEBI:16509
C1=CC(=O)C=CC1=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Very toxic to aquatic life.
Toxic if swallowed.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-405-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only